Learn More
Essendant ADHESIVEEPOXY ULT25ML
Supplier:  Essendant GOR113442
Encompass_Preferred
Our strongest epoxy this Ultimate formula has a 4 250 PSI bond strength and sets in just 10 minutes Gorilla Epoxy Ultimate is also waterproof and versatile creating a long-lasting permanent bond to metal plastics concrete ceramics PVC fiberglass and more (Not recommended for use on polyethylene polypropylene or similar plastics Bonded wood materials should not be submerged or soaked in water ) Adhesive Type- Epoxy Product Packaging- Syringe Washable/Removable- No Color on Application- Black
Catalog No. 50-000-34864
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.