Learn More
Essendant ADHESIVEKRAZYGLUADVNCE
Supplier:  Essendant EPIKG48348CO
Encompass_Preferred
Advanced formula provides an extra strong durable shock-resistant bond on ceramic glass metal plastic wood and rubber The precision tip applicator has an extended no-clog tip with an easy-open easy-to-seal hanger cap Dries clear Adhesive Type- Liquid Product Packaging- Tube Washable/Removable- Yes Color on Application- Clear
Catalog No. 50-000-32442
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.