Learn More
Medchemexpress LLC Adipic acid-13C (hexanedioic acid-13C) | 2708283-72-9 | 98.6% | 147.13 | C513CH10O4 | 10 MG

Supplier: Medchemexpress LLC HYW017522S310MG
Adipic acid-13C is a 13C-isotopically labeled adipic acid supplied as a white to off-white solid for use as a stable isotope reference in analytical chemistry, mass spectrometry, tracer and metabolic studies. The material is intended for analytical workflows and includes storage recommendations to preserve isotopic integrity.
- High isotopic labeling for reliable mass spectrometry internal standard performance.
- Purity 98.6% by manufacturer specification.
- White to off-white solid suitable for standard laboratory handling.
- Storage stability: powder at -20 °C (up to 3 years); in solvent store colder to prolong life.
- Available in milligram-scale quantities for analytical and tracer applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.