Learn More
Cellular Engineering Technologies Adipogenic Differentiation Media (450mL)
Supplier:  Cellular Engineering Technologies MR1007
Encompass_Preferred
Human Adipogenic Differentiation Media is designed to support the differentiation of adipose or bone marrow-derived mesenchymal stem cells into adipose or fat-producing cells. Media is compatible with available animal-origin serum, supplemented separately according to the customer preference.Media is shipped with gel packs.
Catalog No. NC2753070