Learn More
Enzo Life Sciences Adiponectin (human), (recombinant) (50 µg)
SDP
Supplier:  Enzo Life Sciences ALX522063C050
Encompass_Preferred
Synonyms: Adipocyte complement related protein of 30kDa, ACRP30, AdipoQ, apM1, GBP28. MW: ~33kDa (SDS-PAGE). UniProt ID: Q15848. Source: Produced in HEK 293 cells. Human adiponectin (aa 16-244) is fused at the N-terminus to a linker peptide (14 aa) and a FLAG-tag. Formulation: Lyophilized. Contains PBS.
Catalog No. 50-210-0500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.