Learn More
AdipoGen Adiponectin (human) ELISA Kit (96 wells)
SDP
Supplier:  AdipoGen AG45A0001YEKKI01
Encompass_Preferred
ELISA Assay. Detects human adiponectin. Does not cross-react with mouse adiponectin, rat adiponectin, human resistin, human RELM-beta or human leptin. Colorimetric assay. Sample Types: Cell Culture Supernatant, Plasma, Serum, Urine. Range: 0.5 to 32ng/ml. Sensitivity: 100pg/ml. Adiponectin is a promising biomarker of insulin resistance and type 2 diabetes mellitus (T2DM) but also as a potential target for management of the metabolic syndrome. It is a very robust marker that is not prone to degradation or acute inflammatory challenges, is present in relatively high concentrations in the peripheral circulation, and can be collected by a variety of methods. Adiponectin exerts anti-atherogenic and anti-inflammatory properties and may be important as a biomarker for obesity-related cardiovascular disease (CVD). Adiponectin serum level was also described as a good biomarker of colorectal adenoma.
Catalog No. 50-205-3305
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.