Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Adiponectin Receptor 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1059
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1-059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1-059 Supplier Invitrogen™ Supplier No. PA1059
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PA1-059 detects human and rat adiponectin receptor 1. PA1-059 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~42 kDa protein corresponding to human adiponectin receptor 1 from transfected hepatocytes. The PA1-059 immunogen is a synthetic peptide corresponding to residues R(20) E A D T V E L A E L G P L L E E K(37) of human Adiponectin Receptor 1.

Adiponectin (also known as 30- kDa adipocyte complement-related protein; Acrp30) is a hormone secreted by adipocytes that acts as an antidiabetic and anti-atherogenic adipokine. Levels of adiponectin in the blood are decreased under conditions of obesity, insulin resistance and type 2 diabetes. Administration of adiponectin causes glucose-lowering effects and ameliorates insulin resistance in mice. Conversely, adiponectin-deficient mice exhibit insulin resistance and diabetes. This insulin-sensitizing effect of adiponectin seems to be mediated by an increase in fatty-acid oxidation through activation of AMP kinase and PPAR-. Here we report the cloning of complementary DNAs encoding adiponectin receptors 1 and 2 (AdipoR1 and AdipoR2) by expression cloning. AdipoR1 is abundantly expressed in skeletal muscle, whereas AdipoR2 is predominantly expressed in the liver. These two adiponectin receptors are predicted to contain seven transmembrane domains, but are structurally and functionally distinct from G-protein-coupled receptors. Expression of AdipoR1/R2 or suppression of AdipoR1/R2 expression by small-interfering RNA supports our conclusion that they serve as receptors for globular and full-length adiponectin, and that they mediate increased AMP kinase and PPAR- ligand activities, as well as fatty-acid oxidation and glucose uptake by adiponectin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Adiponectin Receptor 1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene ADIPOR1
Gene Accession No. Q96A54
Gene Alias 2810031L11Rik; ACDCR1; adiponectin receptor 1; adiponectin receptor protein 1; Adipor1; CGI45; CGI-45; PAQR1; Parq1; Progestin and adipoQ receptor family member 1; Progestin and adipoQ receptor family member I; TESBP1A
Gene Symbols ADIPOR1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues R(20) E A D T V E L A E L G P L L E E K(37) of human Adiponectin Receptor 1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 289036, 51094
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.