Learn More
Carbosynth LLC. Guinea Pig polyclonal Adipophilin antibody
SDP
Supplier:  Carbosynth LLC. 20RAP002
Encompass_Preferred
Adipophilin / ADRP (a member of the PAT family) is a ubiquitous component of lipid droplets. It has been found in milk fat globule membranes and on the surface of lipid droplets in various cultured cell lines (see e.g. Heid et al.; for review see e.g. Targett-Adams et al.); inducible by etomoxir. Enhanced expression of Adipophilin / ADRP is a useful marker for pathologies characterized by increased lipid droplet accumulation. Such diseases include atheroma, steatosis, obesity and certain cases of liposarcoma. It also seems to be a potent marker for atherosclerosis.
Catalog No. 50-125-2879
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.