Learn More
Selleck Chemical LLC AdipoRon
SDP
Supplier:  Selleck Chemical LLC S736510MG
Encompass_Preferred
AdipoRon (SC-396658) is a novel and orally bioavailable adiponectin receptor agonist with KD of 1 8 and 3 1 M for AdipoR1 and AdipoR2 respectively
Catalog No. 50-217-3268
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.