Learn More
Abcam AdipoRon, Adiponectin Receptor Agonist, >98% Purity, 50 mg
Description
- Adiponectin receptor agonist: KD values are 1.8 μM and 3.1 μM for AdipoR1 and AdipoR2, respectively.
- Signaling activator: Activates AMPK and PPARα signaling.
- Biological effects: Shows antiobesogenic, antidiabetic, and cardioprotective effects in vivo.
- Physical properties: MW 428.5 Da, Purity >98%.
- Administration: Orally active.
- Form: Solid.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Adiponectin Receptor |
| Substrate Properties | Chemical Substrate |
| Buffer | 98% 2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide |
| Content And Storage | Store at -20°C. |
| Form | Solid |
| Formulation | 98% 2-(4-benzoylphenoxy)-N-(1-benzylpiperidin-4-yl)acetamide |
| Quantity | 50 mg |
| For Use With (Application) | AMPK and PPARα signaling activation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.