Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety SMS Adjustable Bouffant Cap
SDP

Catalog No. 19120687
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-120-687 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-120-687 Supplier Keystone Safety Supplier No. 110NWSMI24
This item is not returnable. View return policy

Keystone Polypropylene Bouffant Cap keeps hair out of your eyes and away from your work area with sanitary hair covers. It is ideal for food processing, electronic and pharmaceutical manufacturing or general industry.

  • Polypropylene
  • 100% Latex Free
  • Economical Protection
  • Manufactured in the U.S.

Specifications

Product Type Bouffant
Material Polypropylene
Color White
Disposable Single-use
Latex Free Yes
Model 110NWSM
Sterility Non-sterile
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Style Traditional
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.