Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Polypropylene Bouffant Caps
SDP

Catalog No. 19055463
Change view
Click to view available options
Color:
Green
Navy
Orange
Red
White
Yellow
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-055-463 Green
19-055-460 White
19-055-461 -
19-722-000 White
19-722-002 -
19-055-261 Orange
19-055-464 Red
19-055-465 Navy
19-055-466 Yellow
19-055-467 Green
19-068-496 White
19-055-462 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Catalog No. 19-055-463 Supplier Keystone Safety Supplier No. 110NWI1021GREEN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Bouffant caps keep hair in place and help ensure a clean and hygienic work area. Available in a variety of sizes and colors.

  • Polypropylene
  • 100% Latex Free
  • Economical Protection
  • Manufactured in the USA

Specifications

Product Type Bouffant
Material Polypropylene
Color Green
Disposable Single-use
Latex Free Yes
Model 110NWI
Sterility Non-sterile
Certifications/Compliance FFDCA Certified, IES-RP-CC-003.2
Quantity 1000/Cs.
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Style Traditional
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.