Learn More
Keystone Safety SMS Coveralls, CVL-SMS-E
SDP
Catalog No. 19068493
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-068-493 2X-Large
19-068-494 3X-Large
19-068-492 X-Large
19-068-491 Large
19-068-490 Medium
19-068-495 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-068-493 Supplier Keystone Safety Supplier No. CVLSMSE2XLWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy
Two layers of spunbonded polypropylene enhanced with an inner layer of meltblown polypropylene offers barrier protection while keeping wearers cool and comfortable.
  • Soft and durable SMS material
  • Two layers of spunbonded polypropylene enhanced with an inner layer of meltblown polypropylene
  • Barrier protection while keeping the worker cool, comfortable and does not sacrifice mobility
  • Elastic wrists and ankles
  • Lightweight
  • 100% latex free
  • Fluid and tear/puncture resistant
Specifications
Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Collar Type Standard Fold
Latex Free Yes
Model CVL-SMS-E