Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety White Polypropylene Lab Coats, Elastic Wrists
SDP

Catalog No. 19055501
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Cuff Style:
Elastic
No. of Pockets:
3
None
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style No. of Pockets
19-055-501 Medium Elastic 3
19-068-550 Small Elastic None
19-068-551 Medium Elastic None
19-068-552 Large Elastic None
19-068-553 X-Large Elastic None
19-068-554 2X-Large Elastic None
19-068-555 3X-Large Elastic None
19-068-556 4X-Large Elastic None
19-055-502 Large Elastic 3
19-068-557 5X-Large Elastic None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Catalog No. 19-055-501 Supplier Keystone Safety Supplier No. LC3WENWMEDWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cool and comfortable polypropylene lab coats are ideal for nonhazardous areas where a low particulate barrier is needed.

  • Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
  • Non-woven polypropylene material is durable and low linting
  • Ideal for use in food manufacturing, light industrial, and medical environments
  • Full collar, elastic wrists
  • 3-pocket style available
  • Packaged 30 per case
  • Meets ANSI 101-1996

Specifications

Product Type General Purpose Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
No. of Pockets 3
Collar Type Single
Latex Free Yes
Model LC3-WE-NW
Seam Type Serged
Certifications/Compliance ANSI 101-1996
For Use With (Application) Food, Manufacturing, Light Industrial, Medical
Autoclavable Not Autoclavable
No. per Pack 30/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.