Learn More
Keystone Safety White Polypropylene Lab Coats, Elastic Wrists

Description
- Lab coats provide protection against dirt, grime, and non-hazardous dry particulates
- Non-woven polypropylene material is durable and low linting
- Ideal for use in food manufacturing, light industrial, and medical environments
- Full collar, elastic wrists
- 3-pocket style available
- Packaged 30 per case
- Meets ANSI 101-1996
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| No. of Pockets | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.