Learn More
Pyramex Safety Products Adjustable Cooling Vest
Catalog No. 19555029 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
2X-Large to 5X-Large
Medium to X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-029 Medium to X-Large
19-555-030 2X-Large to 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-555-029 Supplier Pyramex Safety Products Supplier No. CV100M
Only null left
Add to Cart
Add to Cart
Pyramex Safety™ Cooling Vest features evaporative PVA panels to keep users cool for up to 8 hours.
  • Adjustable waist band
  • Quilted outer shell is made from soft polyester
  • Zipper front closure
  • Reusable and washable
  • Submerge in cool water 1 to 2 minutes before wearing to activate
  • CV100 Series
Specifications
Material PVA, Polyester
Size Medium to X-Large
Color Gray
Closure Type Zipper