Learn More
Mopec Adjustable Filler With Toe Kick, Standing Height
SDP
Catalog No. 2123672 Shop All Mopec Products
Change view
Click to view available options
Product Type:
Space Filler
Catalog No. Product Type
21-236-72 Space Filler
1 options
Catalog No. 21-236-72 Supplier Mopec Supplier No. LCFFHADJS
Add to Cart
Add to Cart
Mopec’s Casework Line offers durable, modular stainless-steel cabinetry for surgical pathology, autopsy, and lab settings. Built for longevity, easy cleaning, and seamless integration, it supports efficient, organized, and ergonomic workflows in clinical environments.
Construction
  • (1) Standing height base filler with toekick
Materials
  • 304 stainless steel construction with a polished No. 4 finish
Dimensions
  • LCFFH-ADLC: 18.0 in. L x 1.0 in. W x 34.6 in. H
Specifications
Product Type Space Filler