Learn More
Electron Microscopy Sciences Adjustable Tube Rack Easel
SDP
Supplier:  Electron Microscopy Sciences 7098702
Encompass_Preferred
Angle tube racks at 25 degree, 35 degree or 45 degree to maximize visual and pipetting comfort. Support base grooves allow user to quickly switch between viewing angles. Easel is designed to rest directly on benchtop at all angles to increase overall tube rack stability. Simply pick an angle, place the easel in the base and start pipetting.
Catalog No. 50-192-7375
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.