Learn More
TARGETMOL CHEMICALS INC ADL-5747 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T29663
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. ADL-5747 (ADL-5747 (free base)) (free base) is a delta opioid receptor agonist used in the treatment of neurological disorders skin and musculoskeletal disorders and may be used in the study of postherpetic neuralgia knee osteoarthritis and pain. purity: 99%
Catalog No. 50-258-0450
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.