Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ QuantiGene™ Plex Human ADMETOX Panel, 80-plex

Catalog No. 502417803
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Plate
10 Plates
3 Plates
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-241-7803 10 Plates
50-241-7811 1 Plate
50-241-7809 3 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-241-7803 Supplier Invitrogen™ Supplier No. QGP380ADMETOX
Add to Cart
Add to Cart
Request a Quote

The QuantiGene Plex Human ADMETOX Panel, 80-plex, enables the measurement of gene expression of all relevant ADME (absorption, distribution, metabolism, and excretion) and toxicological biomarkers through the analysis of 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification.

The QuantiGene Plex Human ADMETOX Panel, 80-plex, enables the measurement of gene expression of all relevant ADME (absorption, distribution, metabolism, and excretion) and toxicological biomarkers through the analysis of 80 RNA targets in a single well using Luminex xMAP technology in combination with branched DNA signal amplification. Analyze 80 genetic targets implicated in drug metabolism, including enzymes, transporters, and CYPs, as well as toxicology and apoptosis for efficient profiling of your drug discovery and basic research samples.

QuantiGene Plex panels are extensively tested for target combinability, interference, and cross-reactivity to provide the highest level of validation and precision. This kit is not compatible with the MAGPIX instrument.

This panel is used in combination with the QuantiGene Plex Assay Kit (Cat. No. QP1013), which contains all buffers, plates, and detection reagents necessary to perform the assay in 96-well plates. For detailed information, see create custom research panels

  • Rapid turnaround for this catalog panel for fast delivery
  • Targets: ABCA1, ABCB1 , ABCB11, ABCC1, ABCC2, ABCC3 , ABCG2 , ACTB, AOX1, B2M, BAX, BCL2, CASP3,CASP9, CES1, CYP1A1, CYP1A2, CYP2A6, CYP2B6, CYP2C19, CYP2C8, CYP2C9, CYP2D6, CYP2E1, CYP2J2, CYP3A4, CYP3A5, CYP3A7, DPYD, FMO1, FMO3, FMO5, GAPDH, GSTA2, GSTM1, GSTP1, GSTT1, GUSB, HMGB1, HMGCS2, HNF4A, HPRT1, NAT1, NAT2, PGK1, POLR2A, PPIB, RIPK1, RXRA, SLC10A1, SLC15A1, SLC15A2, SLC16A1, SLC22A1, SLC22A2, SLC22A6, SLC22A7, SLC2A1, SLC3A1, SLCO1B1, SLCO1B3, SLCO2B1, SULT1A1, SULT2A1, SULT2B1, TBP, TFRC, TP53, TPMT, UGT1A1, UGT1A10, UGT1A3, UGT1A4, UGT1A6, UGT1A9, UGT2B15, UGT2B17, UGT2B4, UGT2B7, XDH

    Panel and signal amplification technology

    QuantiGene Plex assays are hybridization-based assays for the direct quantitation of RNA from cell lysates, tissue homogenates, or purified RNA. They enable the measurement of multiple targets in the same well via multiplexing.

    QuantiGene Plex panels contain two components: a target-specific probe set and magnetic capture beads. The target-specific probes are used to capture the RNA of interest, and branched DNA technology then amplifies the fluorescent signals, which are read with a Luminex xMAP instrument. The fluorescent signal is directly proportional to the amount of target mRNA present in the sample. This allows for (relative) quantification of gene expression and with this specific panel enables human immune profiling. Detailed and step-by-step data analysis is facilitated by the Trusted Sustainability Partner
    Suppliers who meet our established requirements for corporate social responsibility

    Learn More" alt="TRUSTED_SUSTAINABILITY" title="TRUSTED_SUSTAINABILITY" />

    Specifications

    Format 96-well
    For Use With (Application) QuantiGene Gene Expression Assay
    For Use With (Equipment) Luminex Instrument
    Content And Storage

    Target-specific probe set and magnetic capture beads (premixed and ready to use).

    Store probe set and capture beads at 4°C.

    Do not freeze the capture beads, as they can be damaged if frozen.

    Product Line QuantiGene™ Plex
    Product Type Multiplex Panel
    Quantity 10 Plates
    Shipping Condition Wet Ice
    Sample Type Purified RNA, cell/blood/FFPE lysates, tissue homogenates
    Detection Method Fluorescence
    Research Area Toxicology
    Species Human
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.