Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Admittance Signs

Catalog No. 19103667 Shop All Brady Corporation Products
Change view
Click to view available options
Material:
Aluminum
Fiberglass, Polyester
Plastic
Color:
Blue on White
Red on White
Height (English):
7 in.
10 in.
Displayed Text:
AUTHORIZED PERSONNEL ONLY
Notice No Entry Unless Authorized
Restricted Area Authorized Employees Only
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Color Height (English) Displayed Text
19-103-667 Plastic Red on White 10 in. Restricted Area Authorized Employees Only
17-046-6 Fiberglass, Polyester Red on White 10 in. AUTHORIZED PERSONNEL ONLY
19-016593 Fiberglass, Polyester Red on White 10 in. Restricted Area Authorized Employees Only
19-017711 Fiberglass, Polyester Blue on White 7 in. AUTHORIZED PERSONNEL ONLY
19-807-064 Aluminum Blue on White 10 in. Notice No Entry Unless Authorized
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-103-667 Supplier Brady™ Supplier No. 22178
Add to Cart
Add to Cart

Alert employees to potential hazards, safety warning and cautionary notices

  • Indicate and identify areas of access
  • Restrict access to property
  • Available in Fiberglass reinforced polyester, aluminum, plastic and polyester
  • Available in variety of sizes

Specifications

Material Plastic
Height (English) 10 in.
Height (Metric) 25.4 cm
Width (English) 14 in.
Width (Metric) 35.5 cm
Displayed Text Restricted Area Authorized Employees Only
Product Type Admittance Sign
Color Red on White

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.