Learn More
Cayman Chemical Adnopoa tapyla Lupnon 25mg
SDP
Supplier:  Cayman Chemical 3533525 MG
Encompass_Preferred
A triterpenoid; inhibits HIV-1 reverse transcriptase (IC50 = 2.1 μM); active against the W2 and D6 strains of P. falciparum (IC50s = 208.1 and 394.7 ng/ml, respectively); induces melanogenesis in (EC50 = 0.35 µM), and inhibits the growth of, B16 2F2 cells in vitro (IC50 = 25.4 µM); topical application reduces TPA-induced EAr edema in mice at 1 mg/ear; inhibits acetic acid-induced writhing in mice at 100 mg/kg
Catalog No. 50-291-3990