Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADNP Polyclonal Antibody, Invitrogen™

Catalog No. PIPA547792
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547792 Supplier Thermo Scientific Supplier No. PA547792

Goat Polyclonal Antibody

In direct ELISAs, approximately 30% cross-reactivity with recombinant human ADNP is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

ADNP Polyclonal antibody specifically detects ADNP in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADNP
Applications Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Adnp
Gene Accession No. Q9JKL8, Q9Z103
Gene Alias AA589558; activity dependent neuroprotector homeobox; activity-dependent neuroprotective protein; activity-dependent neuroprotector; activity-dependent neuroprotector homeobox; activity-dependent neuroprotector homeobox protein; Adnp; ADNP homeobox 1; ADNP1; HVDAS; KIAA0784; mKIAA0784; MRD28; NAP peptide
Gene Symbols Adnp
Host Species Goat
Immunogen E. coli-derived recombinant mouse ADNP Leu941-Ala1102
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11538, 64622
Target Species Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.