Learn More
Invitrogen™ ADNP Recombinant Rabbit Monoclonal Antibody (16H17L68)
Description
This antibody is predicted to react with Rat, Bovine.
Specifications
Specifications
| Antigen | ADNP |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | 16H17L68 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2-7.4 |
| Gene | Adnp |
| Gene Accession No. | Q9H2P0, Q9Z103 |
| Gene Alias | AA589558; activity dependent neuroprotector homeobox; activity-dependent neuroprotective protein; activity-dependent neuroprotector; activity-dependent neuroprotector homeobox; activity-dependent neuroprotector homeobox protein; Adnp; ADNP homeobox 1; ADNP1; HVDAS; KIAA0784; mKIAA0784; MRD28; NAP peptide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.