Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Adrenocorticotrophic Hormone (ACTH) Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. ms452pabx Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
7 mL
Form:
Concentrated
Prediluted
Concentration:
1mg/mL
200μg/mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
MS452PABX 200 μL Concentrated 1mg/mL
MS452P 1 mL Concentrated 200μg/mL
MS452P0 100 μL Concentrated 200μg/mL
MS452P1ABX 100 μL Concentrated 1mg/mL
MS452P1 500 μL Concentrated 200μg/mL
MS452R1 1 mL Prediluted -
MS452R7 7 mL Prediluted -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MS452PABX Supplier Epredia Supplier No. MS452PABX

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Adrenocorticotrophic Hormone (ACTH) Ab-1, Mouse Monoclonal Antibody.

Host Species: Mouse

Clone: AH26

Isotype: IgG1

Species Reactivity: Human. It shows a broad species reactivity.

Epitope: aa 1-24

Immunogen: A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH

Positive Control: Normal pituitary gland or pituitary adenoma

Cellular Localization: Cytoplasmic Ab-1 is excellent for staining of routine formalin-fixed, paraffin-embedded tissues. It labels corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas. NeoMarkers' Ab-1 and Ab-2 may provide a pair for ELISA

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Antigen Adrenocorticotrophic Hormone (ACTH) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone AH26
Concentration 1mg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen A synthetic peptide corresponding to aa 1-24 from the N-terminal of human ACTH
Quantity 200 μL
Regulatory Status IVD
Research Discipline Steroids Hormones and Growth Factors
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.