Learn More
Diagnostic Biosystems ADRENOCORTICOTROPIC HORMONE Adrenocorticotropic Hormone (ACTH) - (AH26)
Supplier:  Diagnostic Biosystems MOB24405
Encompass_Preferred
This antibody is specific to Synacthen (aa1-24 of ACTH); does not react with CLIP (aa17-39 of ACTH). POMC (peo-opiomelanocortin or corticotropinlipotropin) is a 267 amino acid polypeptide hormone precursor that goes through extensive, tissue-specific posttranslational processing by convertases. This antibody stains corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas.
Catalog No. 50-238-5502
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.