Learn More
CPC Scientific H-Tyr-Arg-Gln-Ser-Met-Asn-Asn-Phe-Gln-Gly-Leu-Arg-Ser-Phe-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Gly-Val-Ala-Pro-Arg-Ser-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM012B
Encompass
Participating hormone in blood pressure control that also elicits a potent and long lasting hypotensive effect.ONE-LETTER SEQUENCE: YRQSMNNFQGLRSFGCRFGTCTVQKLAHQIYQFTDKDKDGVAPRSKISPQGY-NH2 (Cys16 and 21 bridge)MOLECULAR FORMULA: C262H403N79O76S3MOLECULAR WEIGHT:5971.8STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [912862-96-5], SYNONYMS: ADM (porcine), AdrenomedullinRESEARCH AREA: Cardiovascular
Catalog No. 50-210-8638
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.