Learn More
CPC Scientific H-Ser-Thr-Gly-Cys-Arg-Phe-Gly-Thr-Cys-Thr-Met-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-Gln-Phe-Thr-Asp-Lys-Asp-Lys-Asp-Gly-Met-Ala-Pro-Arg-Asn-Lys-Ile-Ser-Pro-Gln-Gly-Tyr-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific ADRM010B
Encompass
C-terminal fragment found to induce dose-dependent and endothelium-independent vasodilatation on arterial mesenteric vasculator, though inactive on the venous side of tested rat perfused mesentric bed. This peptide thus shares similar properties to those of CGRP (human)ONE-LETTER SEQUENCE: STGCRFGTCTMQKLAHQIYQFTDKDKDGMAPRNKISPQGY-NH2 (Cys4 and 9 bridge)MOLECULAR FORMULA: C194H304N58O59S4MOLECULAR WEIGHT:4521.17STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [163648-32-6], SYNONYMS: Adrenomedullin (11-50) (rat)RESEARCH AREA: CardiovascularREFERENCES: N.Berthiaume et al., Can. J. Physiol. Pharmacol., 73, 1080 (1995); A.M.Elhawary et al., Br. J. Pharmacol., 115, 1133 (1995)
Catalog No. 50-210-8634
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.