Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Excelta™ Adson Tissue Forceps with 1 x 2 Teeth

Catalog No. 16050139 Shop All Excelta Corporation Products
Change view
Click to view available options
For Use With (Application):
Handling large soft tissue
Handling medium soft tissue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
16-050-139 Handling large soft tissue
16-050-134 Handling medium soft tissue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-050-139 Supplier Excelta™ Supplier No. F6SE

missing translation for 'validMainframeMsg'

Adson Tissue Forceps with 1 x 2 Teeth used for holding and manipulating delicate tissues.

  • Handles are serrated for better gripping
  • Satin finish reduces glare
  • 1 x 2 teeth configuration allows for absolute gripping of soft tissue
  • Made of surgical stainless steel

Specifications

Product Type Forceps
Material Stainless Steel
Finish Satin
For Use With (Application) Handling large soft tissue
Includes 1 x 2 Teeth Forceps
Quantity 1 Ea.
Style Adson
Teeth Count 1 x 2
Tip Style With Teeth
Length (English) 4.5 in.
Length (Metric) 113 mm
Width (English) 0.098 in.
Width (Metric) 2.54 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.