Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec™ Adult Body Bag with 8 Handles, 16 mil, Black
SDP

Mopec™ partners with American Professional Manufacturing™ to provide these sturdy and durable body bags.

Supplier:  Mopec™ BE801EBKXL

 View more versions of this product

Catalog No. 22-445-654


Only null left
Add to Cart

Description

Description

  • Custom-made, lockable slider for easy opening and closing
  • Made of PVC laminated polyester
  • HD nylon zipper
  • 100% sewn with nylon thread
  • Dielectrically sealed seams
  • Available with center or envelope zipper
  • 8 handles secured by heavy-duty steel rivets
  • Leak resistance
Specifications

Specifications

>300 lb.
Black
5 Bags/Cs.
16 mil
94 in.
48 in.
Body Bag
Envelope Zipper
PVC
X-Large
94 x 48 in.
238.76 cm
121.92 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.