Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec™ Adult Body Bag with Envelope Zipper, 4 to 8 mil

Mopec™ partners with American Professional Manufacturing™ to provide these sturdy and durable body bags.
Supplier: Mopec™ BE802EY
Description
- Custom-made, lockable slider for easy opening and closing
- Dielectrically sealed seams
- Available with center or envelope zipper
- White or yellow color
- Leak resistance
Specifications
200 lb. | |
Yellow | |
12 Bags/Cs. | |
8 mil | |
94 in. | |
36 in. | |
Body Bag |
Envelope Zipper | |
PVC | |
Large | |
94 x 36 in. | |
238.76 cm | |
91.44 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction