Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Adult Denture Model
SCI_ED

Catalog No. S24452 Shop All Eisco Products
Change view
Click to view available options
Dimensions (L x W x H):
13 x 13 x 18 cm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
S24452 13 x 13 x 18 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24452 Supplier Eisco™ Supplier No. AM0230
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Detailed exposure of upper and lower jaw structures.

  • Details tooth roots, spongiosa, vessels and nerves
  • Movable lower jaw with help of spring action

Specifications

Dimensions (L x W x H) 13 x 13 x 18 cm
For Use With (Application) Depicts adult upper and lower jaw with tooth roots, spongiosa, vessels, nerves
Material Plastic
Weight (Metric) 350 g
Product Type Model, Denture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.