Learn More
Onsite Testing Specialists Inc ADULTER-SKREEN Urine Validity Controls
Supplier:  Onsite Testing Specialists Inc 20300002
Encompass_Preferred
ADULTER-SKREEN liquid urine controls are designed to monitor the performance of lateral flow strips or other validity test reagents. Designed to match most adulterant devices on the market, with formulations updated as market changes and new devices become available.
Includes Negative and Positive levels to test for Creatinine, Nitrite, Glutaraldehyde, pH, Specific Gravity, and two different Oxidants (PCC and Peroxide). Refer to package insert for target levels. The ADULTER-SKREEN controls are not CE marked for sale in the European Union.
Sold in ready to use liquid 20ml kits, including one POSTIVE level and one NEGATIVE level.
Catalog No. NC1646075