Learn More
Mitegen Ethane Cryostat Temperature Controller, Advanced Regulation for Liquid Ethane, 100/240V Power, For Reproducible Vitrification
SDP
Supplier:  Mitegen MLTCCS001TC
Encompass_Preferred
This advanced Ethane Cryostat Temperature Controller is designed to precisely regulate the temperature of liquid ethane within a compatible cryostat, ensuring highly reproducible vitrification of biological and material samples for cryo-electron microscopy (cryo-EM). It works synergistically with the Ethane Cryostat (SKU ML-TCCS-001-CD) to maintain optimal freezing conditions, preventing ethane solidification or premature evaporation. This controller is vital for achieving consistent, high-quality sample preparation, which is fundamental for obtaining high-resolution structural data. Power supply rates for input power 100-240VAC 50-60Hz. Supplied with a power cord, this unit integrates seamlessly into various laboratory setups. Its precise temperature regulation enhances experimental control and reliability, minimizing variables during critical plunge-freezing steps.
Catalog No. NC2295126
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.