Description
- Model includes injectable metacarpal, digital and thumb veins on dorsal surface, one pint of simulated blood, two I.V. bags, butterfly set, 3 cc syringe, and 22 gauge needle
- Soft, realistic, flexible fingers are separately molded
- Flexible wrist for easy, accurate movement
- Skin rolls when veins are palpated
- Perform hundreds of injections and practices before replacing vein and skin
Specifications
Specifications
| Material | Silicone |
| Gender | Male, Female |
| Product Type | Training Model |
| Grade Level | Higher Ed |