Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Advance Tabco™ Premium Series Stainless-Steel Work Tables

Catalog No. 19999595 Shop All Advance Tabco Products
Change view
Click to view available options
Dimensions (L x W x H):
24 x 24 x 35 in. (60.96 x 60.96 x 88.9 cm)
30 x 24 x 35 in. (76.2 x 60.96 x 88.9 cm)
30 x 24 x 35.5 in. (76.2 x 60.96 x 90.17 cm)
36 x 24 x 35.5 in. (91.44 x 60.96 x 90.17 cm)
36 x 24 x 36 in. (0.91 x 76.2 x 91.4 cm)
48 x 24 x 35.5 in. (121.92 x 60.96 x 90.17 cm)
48 x 30 x 35.5 in. (121.92 x 76.2 x 90.17 cm)
60 x 24 x 35.5 in. (152.4 x 60.96 x 90.17 cm)
60 x 30 x 35.5 in. (152.4 x 76.2 x 90.17 cm)
72 x 24 x 32 in. (182.88 x 60.96 x 88.9 cm)
72 x 24 x 35.5 in. (182.88 x 60.96 x 90.17 cm)
72 x 30 x 35.5 in. (182.88 x 76.2 x 90.17 cm)
84 x 30 x 35.5 in. (213.36 x 76.2 x 90.17 cm)
96 x 24 x 35.5 in. (243 x 60.96 x 90.17 cm)
Includes:
Three hat channels
Two hat channels
Weight (English):
25 lb.
42 lb.
49 lb.
53 lb.
64 lb.
72 lb.
79 lb.
85 lb.
89 lb.
92 lb.
93 lb.
107 lb.
109 lb.
113 lb.
126 lb.
700 lb.
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H) Includes Weight (English)
19-999-595 96 x 24 x 35.5 in. (243 x 60.96 x 90.17 cm) Two hat channels 700 lb.
19-033-551A 30 x 24 x 35.5 in. (76.2 x 60.96 x 90.17 cm) Two hat channels 49 lb.
19-033-551C 36 x 24 x 35.5 in. (91.44 x 60.96 x 90.17 cm) Two hat channels 64 lb.
19-033-551D 48 x 24 x 35.5 in. (121.92 x 60.96 x 90.17 cm) Two hat channels 79 lb.
19-03-3-551E 60 x 24 x 35.5 in. (152.4 x 60.96 x 90.17 cm) Two hat channels 92 lb.
19-033-551F 72 x 24 x 35.5 in. (182.88 x 60.96 x 90.17 cm) Two hat channels 109 lb.
19-033-552E 60 x 30 x 35.5 in. (152.4 x 76.2 x 90.17 cm) Three hat channels 107 lb.
19-033-552D 48 x 30 x 35.5 in. (121.92 x 76.2 x 90.17 cm) Three hat channels 89 lb.
19-033-552F 72 x 30 x 35.5 in. (182.88 x 76.2 x 90.17 cm) Three hat channels 126 lb.
19-999-597 24 x 24 x 35 in. (60.96 x 60.96 x 88.9 cm) Two hat channels 25 lb.
19-999-596 30 x 24 x 35 in. (76.2 x 60.96 x 88.9 cm) Two hat channels 42 lb.
19-999-601 72 x 24 x 32 in. (182.88 x 60.96 x 88.9 cm) Two hat channels 85 lb.
19-033-554D 48 x 30 x 35.5 in. (121.92 x 76.2 x 90.17 cm) Three hat channels 72 lb.
19-033-554E 60 x 30 x 35.5 in. (152.4 x 76.2 x 90.17 cm) Three hat channels 85 lb.
19-033-554G 84 x 30 x 35.5 in. (213.36 x 76.2 x 90.17 cm) Three hat channels 113 lb.
19-033-554F 72 x 30 x 35.5 in. (182.88 x 76.2 x 90.17 cm) Three hat channels 93 lb.
19-999-598 36 x 24 x 36 in. (0.91 x 76.2 x 91.4 cm) Two hat channels 53 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 19-999-595 Supplier Advance Tabco™ Supplier No. SS248
Add to Cart
Add to Cart
Request a Quote

Wide, adjustable, and with a turned up edge for wall protection, the Advance Tabco Stainless-Steel Work Table is made for, and can take, heavy use in nearly all environments.

  • Top is furnished with 4.12cm (1.625 in.) sanitary rolled rim edges on front and back and square side edges
  • To reinforce and maintain level working surface, tables are supplied with hat channels
  • Pre-engineered welded angle adapters insure ease of future drawer installation

Model Specific:

SS - Series

  • Aluminum die cast leg-to-shelf clamp secures shelf to leg eliminating unsightly nuts and bolts. Undershelf is adjustable
  • Top: 14 gauge stainless steel type 304 series
  • Shelf: 18 gauge stainless steel
  • Legs: 4.12cm (1.625 in.) diameter tubular stainless steel. 2.54cm (1 in.) adjustable stainless steel bullet feet. Stainless steel gussets

TSS - Series

  • Front to back Stretchers are welded to legs. Left to right
  • Top: 14 gauge stainless steel type 304 series
  • Stretchers: 4.12cm (1.625 in.) dia. tubular stainless steel. LEGS: 4.12cm (1.625 in.) dia. tubular stainless steel type 304 series
  • 2.54cm (1 in.) adjustable stainless steel bullet feet. Stainless steel gussets. Stretcher bolted to legs and requires assembly

Specifications

Product Type Worktable
Weight (Metric) 317.5 kg
Length (Metric) 243 cm
Height (English) 35.5 in.
Width (Metric) 60.96 cm
Width (English) 24 in.
Certifications/Compliance NSF
Length (English) 96 in.
Height (Metric) 90.17 cm
Weight (English) 700 lb.
Includes Two hat channels
Finish Satin
Shelf Material Stainless Steel
Dimensions (L x W x H) 96 x 24 x 35.5 in. (243 x 60.96 x 90.17 cm)
Material Stainless Steel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.