Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Advanced Crystal Violet Stain
SDP

Catalog No. 23001760 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Volume (Metric):
250 mL
3.7 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-001-760 3.7 L
23-001-796 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-001-760 Supplier Hardy Diagnostics™ Supplier No. C128A
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used as a primary stain in the advanced gram stain procedure. An aqueous solution of crystal violet.

  • Crystal Violet is taken up equally well by both gram-positive and negative bacterial cell walls. A crystal violet-iodine complex is formed within the peptidoglycan layer of the cell wall of each type of bacteria upon addition of iodine
  • The advanced system helps to guard against accidental over-decolorization, thus reducing the possibility of mistaking a gram-positive organism for a gram-negative one

Specifications

Format Liquid
Volume (Metric) 3.7 L
Compatible Cells Gram-negative or Gram-positive bacteria
Label or Dye Dye
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.