Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Advanced Electricity And Magnetism Kit
SDPSCI_ED

Catalog No. S139255 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Class Size:
10 to 40 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S139255 10 to 40 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Prop 65
Catalog No. S139255 Supplier United Scientific™ Supplier No. EMKIT4

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Prop 65

A comprehensive kit for the entire class to use to demonstrate electricity and magnetism.

  • This comprehensive 47-piece kit introduces students to the basic principles of electricity and magnetism.
  • Includes materials for studying static electric charges, magnetic attraction and repulsion, magnetic fields, electrical circuitry, voltage, current and resistance, and series and parallel connection.
  • Electromagnetism topics include the force in a magnetic field and electric motor and generator basics.
  • Light emitting diodes, a solar cell, a reed switch, a buzzer, and an electric bell are provided for further investigations.
  • The components are mounted on sturdy molded bases and connections are made with reliable spring terminals.
  • An activity guide details the experiments.
  • Requires three AA batteries (not included).
  • Kit includes DC ammeter (1), DC voltmeter (1), battery holders, AA size (3), knife switches, SPST (2), knife switch, SPDT (1), lamp holders (3), miniature incandescent lamps (8), resistor board (5Ω,10Ω,15Ω, “Unknown”) (1), slide Rheostat (20Ω, 2A) (1), wire resistance board (1), conductivity test board (1), bar magnet (1), U-shaped magnet (1), ring magnets (3), ring magnet carriage (1), compass (1), magnetic field visualizer (1), electric bell (1), electromagnet (1), electromagnetic force device (1), motor model (1), DC Miniature motor (1), hand generator (1), LED board with 2 LEDs (1), solar cell (1), buzzer (1), magnetic reed switch (1), set of electrostatic test objects (1), set of 16 connecting cords (1), set of electrodes and spares (1).

Specifications

Grade 5+
Includes 25 Activities plus a guide
Class Size 10 to 40 Students
Product Type Electricity and Magnetism Kit

Warning: Cancer and reproductive harm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.