Learn More
Description
- Enhanced with a moisturizer and antioxidant blend.
- Gentle formulation clings to hands for mess-free sanitizing.
- Contains vitamin B3 and is free of fragrance, dyes, parabens, phthalates, sulfates and harsh preservatives.
- Made with our high-quality ethyl alcohol, produced in the USA.
- Manufactured according to strict FDA safety and quality standards.
- Refill bottles are easily recyclable to align with your sustainability standards.
- Formula meets USDA Certified Biobased Product Standards for hand cleaners, ECOLOGO UL 2783, and Cradle to Cradle Certified
- 1200 mL refill for the PURELL ES10 Hand Sanitizer Automatic Dispenser (sold separately) bottle is made with recyclable plastic components.
- Collar and alkaline AA battery easily pull apart for easy recyclability and waste reduction.
- Case Includes: 2-PURELL Advanced Hand Sanitizer ULTRA NOURISHING Foam, 1200 mL Refills for the ES10 Automatic Hand Sanitizer Dispenser.
- ES10 dispensers (8324-E1, 8320-E1, 8328-E1) sold separately.
Specifications
Specifications
| Product Type | Hand Sanitizer |
| Color | Clear |
| Form | Foam |
| Size | 1200 mL |
| Solution Type | Hand Sanitizer Ultra Nourishing Foam |
| Volume (Metric) | 1200 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

