Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Purell™ Advanced Hand Sanitizer Gel
SDP

Catalog No. 19043732 Shop All Gojo Industries, Inc. Products
Change view
Click to view available options
Size:
2 L
4 fl. oz.
8 fl. oz.
800 ml
Description:
Bag for Bag-in-Box Dispenser
Economy Size Pump Bottle
Portable Flip Cap Bottle
Table Top Pump Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Description
19-043-732 2 L Economy Size Pump Bottle
19-053-372 4 fl. oz. Portable Flip Cap Bottle
19-057-585 800 ml Bag for Bag-in-Box Dispenser
19-085159 8 fl. oz. Table Top Pump Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-043-732 Supplier Gojo Industries Inc Supplier No. 962504
This item is not returnable. View return policy

Kills more than 99.99% of most common germs that may cause illness.

  • Refreshing gel
  • Kills more than 99.99% of most common germs that may cause illness
  • Outperforms other hand sanitizers ounce-for-ounce
  • Clinically-proven to maintain skin health
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Gel
Description Economy Size Pump Bottle
Size 2 L
Volume (Metric) 2 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.