Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Filter Screw Cap
GREENER_CHOICE

Catalog No. 07000171 Shop All Greiner Bio-One Products
Change view
Click to view available options
Culture Area:
175 cm2
75 cm2
Volume (Metric) Working:
20 to 45 mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Volume (Metric) Working
07-000-171 175 cm2 20 to 45 mL
07-000-655 75 cm2 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-171 Supplier Greiner Bio-One Supplier No. 660975
Add to Cart
Add to Cart

For maintenance of adherent cells or suspension cell culture. Greiner Bio-One Advanced TC Treated Suspension Cell Culture Flasks with Filter Screw Cap provides optimal conditions for cultivation of sensitive and fastidious cells.

  • Made of high-grade polystyrene
  • Free of detectable DNases, RNases, human DNA, endotoxins and are non-cytotoxic
  • Stacking rim on the top of the flasks ensures firm standing and easy stackability in the incubator
  • Printed graduation for easier filling on both sides
  • Provides homogeneous and optimized cell growth
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens; Contents non-cytotoxic
Closure Color Blue
Closure Material HDPE
Capacity (Metric) 550 mL
Material Polystyrene
Neck Style Canted
No. per Case 50
No. per Pack 5
Surface Coating Tissue Culture Treated
Graduated Yes
Sterility Sterile
Closure Type with filter screw cap
Culture Area 175 cm2
Volume (Metric) Working 20 to 45 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.