Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Advanced Thermostat Kit
SDPSCI_ED

Catalog No. S83793 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
1 Ea.
5 Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S83793 5 Pk.
S83792 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S83793 Supplier United Scientific™ Supplier No. UNADTHK02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Observe energy transfer and conversion and investigate the systems that keep homes comfortable and safe with this advanced thermostat kit.

This apparatus helps students to observe energy transfer and conversion and investigate the systems that keep homes comfortable and safe. The suggested activities below will: explore the relationship between parallel circuits and current; understand how thermostats and circuit breakers operate; and compare the energy efficiency of LEDs to incandescent bulbs.

Components:
  • 6V DC Connections
  • Multimeter Connections
  • Bulb Sockets (5)
  • Bimetallic Bar Switch
  • On/off Switches (5)
  • 6V Incandescent Bulbs (5)
  • LED Bulbs (5)
  • Candle
  • Multimeter
  • Additional Supplies Needed: 6V DC Power Supply and Stopwatch

Specifications

Grade 6 to 14
Age 12 to 20
For Use With (Application) Teaching about Thermostats
Includes Advanced thermostat apparatus, 5 incandescent bulbs, 5 LED bulbs, Multimeter, Candle (five of each component set), Activity Guide
Quantity 5 Pk.
Product Type Physics Demonstration
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.