Learn More
United Scientific™ AdvancedGen Pendulum Set
SDPSCI_ED
Catalog No. S00338 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
9 to 12
Catalog No. Grade
S00338 9 to 12
1 options
This item is not returnable. View return policy
Catalog No. S00338 Supplier United Scientific™ Supplier No. UNCPNS01
Add to Cart
Add to Cart
This item is not returnable. View return policy
The AdvancedGen Pendulum Set includes a set of twelve 25 mm drilled pendulum balls in six different materials, a triple pendulum clamp, and suspension cord.
  • Bring your physics equipment into the 21st century with this complete pendulum set.
  • The simple pendulum includes a set of twelve 25 mm drilled pendulum balls in six different materials, a triple pendulum clamp, and suspension cord.
    • The balls are counter sunk to the center to allow for suspension at the center of mass.
  • The period dependence and energy in both linear and conical motion can be explored.
  • The physical pendulum consists of a 60 cm aluminum bar with a heavy moveable steel mass and a knife-edge suspension supported by a table clamp.
  • The period dependence on the mass position can be explored and compared with calculated moments of inertia to verify the theoretical formula.
  • The Wilberforce pendulum consists of a heavy cylindrical steel bob with small adjustment masses hanging from a long spiral spring.
  • It can oscillate in two different but coupled modes- linear oscillations (up and down) and torsional oscillations (spinning).
  • The kinetic energy flows back and forth between these modes.
  • The exchange is only complete when the weights are adjusted so that the periods of the two modes are exactly equal.
  • The simple geometry allows the moment of inertia in the spinning mode to be calculated for theoretical comparison of the two periods.
  • The period measurements of each type of pendulum are easily made using a motion sensor.
Specifications
Material Metal
Grade 9 to 12
Product Type Pendulum