Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ Advantage™ 1000 CBA-RCA Gas Masks

Catalog No. 19042200 Shop All MSA Products
Change view
Click to view available options
Format:
Medium
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
19-042-200 Medium
19-042-201 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-042-200 Supplier MSA™ Supplier No. 813859
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Fully elastic, six-point head harness that permits easy donning, doffing and adjustment without pulling hair and features a speaking diaphragm for ease of communications; nosecup with two check valves that deflects exhaled air away from the one-piece polyurethane lens, preventing fogging; filter canister; identification tag.

Clean, simplified respirator is up to 40% lighter than conventional full-face respirators

  • Uses single low-profile filter canister with a bayonet-style attachment
  • Dual-canister mounts permit weapon sighting from either shoulder
  • Super-soft, lightweight facepiece construction contributes to snug, comfortable fit
  • Overall size is compatible with most visored police helmets
  • Flexible lens is bonded to the chemical-resistant Hycar™ facepiece as part of a wraparound design that minimizes wasted space and offers a wide field of vision
  • Canister is also effective against variety of biological agents as well as a list of chemical warfare agents including GA, GB (Sarin), GD, VX, Mustard and Lewisite
  • Tested by MSA and independent laboratories using protocols recommended by the Chemical Agent Safety and Health Policy Action Committee (CASHPAC)
  • Filters provide up to eight hours of protection against CN, CS and oleoresin capsicum (OC) tearing agents

Order Info

Accepts low profile replacement canister Cat. No. 19-042-129 (NIOSH approval TC-14G-0235). Riot control adapter (Cat. No. 19-822-548) and locking gasket (Cat. No.18-999-4137) also available separately.

Certifications

NIOSH-approved protection against tearing agents a-chloroacetophenone (CN) and o-chlorobenzylidine malononitrile (CS), and 99.97% of airborne particulates.

Specifications

Certifications/Compliance NIOSH, CBRN
Connection Type Dual-canister mount
Product Type Gas Mask
Format Medium
For Use With (Application) CBRN incidents, civil disturbances, clandestine labs
Includes Canister, nosecup with two check valve and identification tag

Note: Canister is effective against OC agents and hydrogen cyanide [HCN], but has not been tested or approved by NIOSH in either of these applications. Advantage mask can be used with canister only if supplied port plug is used to seal opposite port.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.