Learn More
Cole-Parmer Advantec GA10047MM Grade GA100 Glass Fiber Filters; 47mm dia; 100/box
Supplier:  Cole-Parmer 0664654
Encompass_Preferred
General purpose; filtering cells and precipitated proteins; air pollution monitoring.
Catalog No. NC1250584