Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chembio Diagnostics, Inc.™ Advin COVID-19 Antigen Test @Home
SDP

Catalog No. 23999122
Change view
Click to view available options
Quantity:
1 Test/Pk.
2 Tests/Pk.
25 Tests/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-999-122 2 Tests/Pk.
23-999-123 1 Test/Pk.
23-999-121 25 Tests/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 23-999-122 Supplier Chembio Diagnostics, Inc.™ Supplier No. 6699901
Add to Cart
Add to Cart

Advin COVID-19 Antigen Test @Home is a lateral flow immunoassay device intended for the qualitative detection of nucleocapsid protein antigens from SARS-CoV-2.

  • Authorized for CLIA waived facilities and non-prescription home use with self-collected anterior nasal (nares) swab samples
  • For individuals aged 14 years or older or with adult-collected anterior nasal (nares) samples from individuals aged 2 years or older
  • Non-invasive
  • Anterior nasal swab specimens; no need to swab deep in the nasal cavity for sample collection
  • Easy to use and offers fast results in just 10 min.
  • Simple 4-step process from sample collection to results

Specifications

CE Marker No
Clia Complexity Waived (EUA, OTC)
Detectable Analytes COVID-19
Detection Method Lateral Flow
DoA Calibrators No
Format Kit
Sample Type Nasal Swab
Type Test Kit
Disposable Single-use
For Use With (Application) At-home COVID-19 Detection
Quantity 2 Tests/Pk.
Sterility Non-sterile
Form Ready-to-Use (RTU)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.