Learn More
Abcam AEC Chromogen Substrate, Liquid, 30 mL
Description
- Highly stable single-component AEC chromogen/substrate working solution
- Form: Liquid
- Constituents: 6% Hydrogen peroxide, 3% 3-amino-9-ethyl carbazole
- Storage: Store at +4°C
- Shipping: Blue Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate | AEC |
| Shipping Condition | Blue Ice |
| Substrate Properties | Chemical Substrate |
| Kit Contents | 30 mL AEC chromogen/substrate working solution (6% Hydrogen peroxide, 3% 3-amino-9-ethyl carbazole) |
| Form | Liquid |
| Product Type | Liquid Substrate |
| Buffer | 6% hydrogen peroxide, 3% 3-amino-9-ethyl carbazole |
| Content And Storage | Store at 4°C. |
| Detection Method | Chromogenic |
| Formulation | 6% Hydrogen peroxide, 3% 3-amino-9-ethyl carbazole |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.