Learn More
R&D Systems™ AEC Chromogen/Substrate
SDP
Catalog No. NB90079773 Shop All Novus Products
Change view
Click to view available options
Quantity:
30 mL
Catalog No. Quantity
NB90079773 30 mL
1 options
Catalog No. NB90079773 Supplier R&D Systems™ Supplier No. NB90079773
Customers require a valid business account associated to their profile to access pricing and ordering options.
A Novus product, part of R&D Systems.
Specifications
Product Line Novus
Quantity 30 mL
For Use With (Application) Stable Chromogen/Substrate Solution to be Used in Conjunction with Peroxidase-Based Immunostaining Systems. Specimens Stained Using AEC Chromogen/Substrate Cannot be Dehydrated in Ethanol and Hence Need to be Mounted in an Aqueous Based Mounting Medium. Staining Procedure 1. Once Sections Have Been Incubated with Peroxidase, Wash Sections with Wash Buffer. 2. Wipe Slides to Remove Excess Buffer. Add Enough Drops of AEC Chromogen/Substrate to Cover Tissue Sections. 3. Incubate for 5- 15 Minutes (at Room Temperature. 4. For Best Results, Observe Reaction Under a Microscope for Signal Development. 5. Once Desired Signal to Noise Ratio is Achieved, Stop Reaction by Washing Slides in DI H2O.)