Learn More
Applied Biosystems™ AEC Solution (IHC)
Catalog No. 5075003
Encompass_Preferred
Change view
Click to view available options
Quantity:
1,000 slides
Catalog No. Quantity
50-750-03 1,000 slides
1 options
This item is not returnable. View return policy
Catalog No. 50-750-03 Supplier Applied Biosystems™ Supplier No. 001122
Add to Cart
Add to Cart
This item is not returnable. View return policy
AEC Solution (IHC)
  • This Single Solution utilizes aminoethyl carbazole (AEC), an alcohol soluble chromogen, and generates a reddish deposit at the reaction site on tissue or cell preparations.
  • Single Solution AEC is suitable for use with manual or automated staining systems, and is provided as a liquid in a single bottle that can be stored at -8°C
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material HRP (Horseradish Peroxidase) Substrate
Quantity 1,000 slides
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.