Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aegeline 98.0+%, TCI America™
SDP

Catalog No. A31705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31705G 5 g
A317025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31705G Supplier TCI America Supplier No. A31705G

Chemical Identifiers

CAS 456-12-2
Molecular Formula C18H19NO3
Molecular Weight (g/mol) 297.35
MDL Number MFCD00048045
InChI Key QRFDENJATPJOKG-UHFFFAOYNA-N
Synonym Aegelin, (E)-N-[2-Hydroxy-2-(4-methoxyphenyl)ethyl]cinnamamide
PubChem CID 15558419
IUPAC Name N-[2-hydroxy-2-(4-methoxyphenyl)ethyl]-3-phenylprop-2-enamide
SMILES COC1=CC=C(C=C1)C(O)CNC(=O)C=CC1=CC=CC=C1

Specifications

Melting Point 180°C
Color White-Yellow
Quantity 5 g
Formula Weight 297.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Aegeline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.